2-Oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Oxo-2,3-dihydrobenzo[d]oxazole-6-carboxylic acid
048556-500MG
048556-1G
101884-360EA
360
USD
101884-360
101884-362
2-Oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylic acid ≥95%
2-Oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylic acid
Hazard: Irritant
Formula:
C₈H₅NO₄ MW: 179.13 g/mol Melting Pt: >340 °C |
MDL Number:
MFCD11179328 CAS Number: 54903-16-1 |
Specification Test Results
Melting point | >340°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...