5-Nitro-6-(trifluoromethyl)-1H-indole ≥95%

Supplier: Matrix Scientific

048555-500MG 048555-1G
101884-356EA 747.69 USD
101884-356 101884-358
5-Nitro-6-(trifluoromethyl)-1H-indole ≥95%
5-Nitro-6-(trifluoromethyl)-1H-indole

Hazard: Irritant

Formula: C₉H₅F₃N₂O₂
Melting Pt: 155…157 °C
MDL Number: MFCD12827792
CAS Number: 1186405-05-9

Order Now

Specification Test Results

Melting point 155-157°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR