4-Nitro-5-(trifluoromethyl)-2-[2-(trimethylsilyl)ethynyl]phenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-nitro-4-[2-(trimethylsilyl)ethynyl]benzotrifluoride

048554-500MG 048554-1G
101884-352EA 556.92 USD
101884-352 101884-354
4-Nitro-5-(trifluoromethyl)-2-[2-(trimethylsilyl)ethynyl]phenylamine ≥95%
4-Nitro-5-(trifluoromethyl)-2-[2-(trimethylsilyl)ethynyl]phenylamine

Hazard: Irritant

Formula: C₁₂H₁₃F₃N₂O₂Si
MW: 302.33 g/mol
Melting Pt: 155…157 °C
MDL Number: MFCD12827775
CAS Number: 1186405-13-9

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR