4-Nitro-2-(trifluoromethyl)phenol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Nitro-2-(trifluoromethyl)benzenol

048553-5G 048553-25G
101884-348EA 207.69 USD
101884-348 101884-350
4-Nitro-2-(trifluoromethyl)phenol ≥95%
4-Nitro-2-(trifluoromethyl)phenol

Hazard: Irritant

Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Melting Pt: 105…107 °C
MDL Number: MFCD11100972
CAS Number: 1548-61-4

Order Now

Specification Test Results

Melting point 105-107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR