N-Methyl-2-(trifluoromethyl)-4-quinazolinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Methylamino)-2-(trifluoromethyl)quinazoline

048552-500MG 048552-1G
101884-344EA 1109.23 USD
101884-344 101884-346
N-Methyl-2-(trifluoromethyl)-4-quinazolinamine ≥95%
N-Methyl-2-(trifluoromethyl)-4-quinazolinamine

Hazard: Irritant

Formula: C₁₀H₈F₃N₃
MW: 227.19 g/mol
Melting Pt: 143…144 °C
MDL Number: MFCD08762128
CAS Number: 929418-48-4

Order Now

Specification Test Results

Melting point 143-144°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR