2-(Methylsulfanyl)-4-piperazino-6-(trifluoromethyl)pyrimidine ≥95%
Supplier: Matrix Scientific
048550-500MG
048550-1G
101884-336EA
747.69
USD
101884-336
101884-338
2-(Methylsulfanyl)-4-piperazino-6-(trifluoromethyl)pyrimidine ≥95%
2-(Methylsulfanyl)-4-piperazino-6-(trifluoromethyl)pyrimidine
Hazard: Irritant
Formula:
C₁₀H₁₃F₃N₄S Melting Pt: 118…119 °C |
MDL Number:
MFCD03618492 CAS Number: 874782-26-0 |
Specification Test Results
Melting point | 118-119°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...