Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-oxo-2,3-dihydrobenzo[d]oxazole-6-carboxylate

048549-500MG 048549-1G
101884-332EA 261.54 USD
101884-332 101884-334
Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylate ≥95%
Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-carboxylate

Hazard: Irritant

Formula: C₉H₇NO₄
MW: 193.16 g/mol
Melting Pt: 216…220 °C
MDL Number: MFCD12827780
CAS Number: 72752-80-8

Order Now

Specification Test Results

Melting point 216-220°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR