Methyl-4-(methylamino)-3-nitrobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(methylamino)-3-nitrobenzoate

048547-500MG 048547-1G 048547-5G
101884-320EA 307.69 USD
101884-320 101884-322 101884-324
Methyl-4-(methylamino)-3-nitrobenzenecarboxylate ≥95%
Methyl-4-(methylamino)-3-nitrobenzenecarboxylate

Hazard: Irritant

Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
Melting Pt: 146…148 °C
MDL Number: MFCD09862450
CAS Number: 36242-50-9

Order Now

Specification Test Results

Melting point 146-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR