Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-iodo-3-methyl-2-[(trifluoroacetyl)amino]benzoate

048545-500MG 048545-1G 048545-5G
101884-312EA 278.46 USD
101884-312 101884-314 101884-316
Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate ≥95%
Methyl-5-iodo-3-methyl-2-[(2,2,2-trifluoroacetyl)amino]benzenecarboxylate

Hazard: Irritant

Formula: C₁₁H₉F₃INO₃
MW: 387.1 g/mol
Melting Pt: 191…193 °C
MDL Number: MFCD12827837
CAS Number: 1186405-10-6

Order Now

Specification Test Results

Melting point 191-193°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR