Methyl-2,3-dichloro-6-quinoxalinecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2,3-dichloroquinoxaline-6-carboxylate

048544-500MG 048544-1G
101884-308EA 216.92 USD
101884-308 101884-310
Methyl-2,3-dichloro-6-quinoxalinecarboxylate ≥95%
Methyl-2,3-dichloro-6-quinoxalinecarboxylate

Hazard: Irritant

Formula: C₁₀H₆Cl₂N₂O₂
MW: 257.08 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD12827814
CAS Number: 108258-54-4

Order Now

Specification Test Results

Melting point 117-119°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR