Methyl-2-(benzyloxy)-5-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 2-(benzyloxy)-5-nitrobenzoate
048543-500MG
048543-1G
048543-5G
101884-302EA
278.46
USD
101884-302
101884-304
101884-306
Methyl-2-(benzyloxy)-5-nitrobenzenecarboxylate ≥95%
Methyl-2-(benzyloxy)-5-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₁₅H₁₃NO₅ MW: 287.27 g/mol Melting Pt: 74…76 °C |
MDL Number:
MFCD12827823 CAS Number: 508211-52-7 |
Specification Test Results
Melting point | 74-76°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...