Methyl-4-amino-3-(benzyloxy)-5-bromobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-amino-3-(benzyloxy)-5-bromobenzoate

048542-500MG 048542-1G
101884-298EA 332.31 USD
101884-298 101884-300
Methyl-4-amino-3-(benzyloxy)-5-bromobenzenecarboxylate ≥95%
Methyl-4-amino-3-(benzyloxy)-5-bromobenzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₄BrNO₃
MW: 336.19 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD12827830
CAS Number: 881909-58-6

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR