Methyl-4-amino-3-(benzyloxy)-5-iodobenzenecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-amino-3-(benzyloxy)-5-iodobenzoate

048541-500MG 048541-1G
101884-294EA 556.92 USD
101884-294 101884-296
Methyl-4-amino-3-(benzyloxy)-5-iodobenzenecarboxylate ≥95%
Methyl-4-amino-3-(benzyloxy)-5-iodobenzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₄INO₃
MW: 383.19 g/mol
Melting Pt: 90…92 °C
MDL Number: MFCD12827829
CAS Number: 1186405-03-7

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR