6-Iodo-8-methyl-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Iodo-8-methyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 1,4-Dihydro-6-iodo-8-methyl-4-oxoquinoline-3-carboxylic acid

048537-500MG 048537-1G
101884-274EA 556.92 USD
101884-274 101884-276
6-Iodo-8-methyl-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid ≥95%
6-Iodo-8-methyl-4-oxo-1,4-dihydro-3-quinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₈INO₃
MW: 329.1 g/mol
Melting Pt: 299 °C (dec.)
MDL Number: MFCD06009652
CAS Number: 773865-48-8

Order Now

Specification Test Results

Melting point 299(dec)°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR