3-Formyl-6-(trifluoromethyl)-1H-indole-5-carbonitrile ≥95%
Supplier: Matrix Scientific
048536-500MG
048536-1G
101884-270EA
921.54
USD
101884-270
101884-272
3-Formyl-6-(trifluoromethyl)-1H-indole-5-carbonitrile ≥95%
3-Formyl-6-(trifluoromethyl)-1H-indole-5-carbonitrile
Hazard: Irritant
Formula:
C₁₁H₅F₃N₂O Melting Pt: 294…296 °C |
MDL Number:
MFCD12827826 CAS Number: 1186404-95-4 |
Specification Test Results
Melting point | 294-296°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...