Ethyl thieno[2,3-b]pyridine-2-carboxylate ≥95%

Supplier: Matrix Scientific

048533-500MG 048533-1G
101884-254EA 130.77 USD
101884-254 101884-256
Ethyl thieno[2,3-b]pyridine-2-carboxylate ≥95%
Ethyl thieno[2,3-b]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₉NO₂S
Melting Pt: 61…63 °C
MDL Number: MFCD07772793
CAS Number: 59944-78-4

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR