Ethyl-4,6-dihydroxy-5-nitronicotinate ≥95%
Supplier: Matrix Scientific
048531-500MG
048531-1G
048531-5G
101884-242EA
230.77
USD
101884-242
101884-244
101884-246
Ethyl-4,6-dihydroxy-5-nitronicotinate ≥95%
Ethyl-4,6-dihydroxy-5-nitronicotinate
Hazard: Irritant
Formula:
C₈H₈N₂O₆ MW: 228.16 g/mol Melting Pt: 243…246 °C |
MDL Number:
MFCD01765414 CAS Number: 6317-97-1 |
Specification Test Results
Melting point | 243-246°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...