Ethyl-2-(benzylamino)-5-chloronicotinate ≥95%

Supplier: Matrix Scientific

048529-500MG 048529-1G
101884-234EA 747.69 USD
101884-234 101884-236
Ethyl-2-(benzylamino)-5-chloronicotinate ≥95%
Ethyl-2-(benzylamino)-5-chloronicotinate

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O₂
MW: 290.75 g/mol
Melting Pt: 40 °C
MDL Number: MFCD12827827
CAS Number: 1186405-01-5

Order Now

Specification Test Results

Melting point 40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR