Ethyl-2-(benzylamino)-5-bromonicotinate ≥95%

Supplier: Matrix Scientific

048528-500MG 048528-1G 048528-5G
101884-228EA 556.92 USD
101884-228 101884-230 101884-232
Ethyl-2-(benzylamino)-5-bromonicotinate ≥95%
Ethyl-2-(benzylamino)-5-bromonicotinate

Hazard: Irritant

Formula: C₁₅H₁₅BrN₂O₂
MW: 335.21 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD12827825
CAS Number: 1186404-92-1

Order Now

Specification Test Results

Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR