Ethyl-2-amino-3-cyano-4,5,6,7-tetrahydro-1-benzothiophene-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-amino-3-cyano-4,5,6,7-tetrahydrobenzo[b]thiophene-4-carboxylate

048527-500MG 048527-1G 048527-5G
101884-222EA 356.92 USD
101884-222 101884-224 101884-226
Ethyl-2-amino-3-cyano-4,5,6,7-tetrahydro-1-benzothiophene-4-carboxylate ≥95%
Ethyl-2-amino-3-cyano-4,5,6,7-tetrahydro-1-benzothiophene-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₂S
Melting Pt: 135…137 °C
MDL Number: MFCD02679242
CAS Number: 175599-83-4

Order Now

Specification Test Results

Melting point 135-137°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR