cis-(6-Ethoxycarbonylmethyl-1-methylpiperazin- 2-yl)acetic acid ethyl ester acetate ≥95%

Supplier: Matrix Scientific

048525-500MG 048525-1G
101884-214EA 556.92 USD
101884-214 101884-216
cis-(6-Ethoxycarbonylmethyl-1-methylpiperazin- 2-yl)acetic acid ethyl ester acetate ≥95%
cis-(6-Ethoxycarbonylmethyl-1-methylpiperazin- 2-yl)acetic acid ethyl ester acetate

Hazard: Irritant

Formula: C₁₅H₂₈N₂O₆
Melting Pt: 88…90 °C
MDL Number: MFCD12827803
CAS Number: 1149624-33-8

Order Now

Specification Test Results

Melting point 88-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR