2,4-Dichloro-5-(chloromethyl)pyrimidine ≥95%
Supplier: Matrix Scientific
048520-500MG
048520-1G
101884-190EA
606.15
USD
101884-190
101884-192
2,4-Dichloro-5-(chloromethyl)pyrimidine ≥95%
2,4-Dichloro-5-(chloromethyl)pyrimidine
Hazard: Irritant/Avoid Skin Contact
Formula:
C₅H₃Cl₃N₂ MW: 197.443 g/mol Melting Pt: 63…64 °C |
MDL Number:
MFCD10697639 CAS Number: 7627-38-5 |
Specification Test Results
Melting point | 63-64°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...