2-Chloro-6-methoxy-4-[2-(trimethylsilyl)ethynyl]-3-pyridinol ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Chloro-3-hydroxy-6-methoxy-4-[2-(trimethylsilyl)ethynyl]pyridine
048517-500MG
048517-1G
101884-180EA
747.69
USD
101884-180
101884-182
2-Chloro-6-methoxy-4-[2-(trimethylsilyl)ethynyl]-3-pyridinol ≥95%
2-Chloro-6-methoxy-4-[2-(trimethylsilyl)ethynyl]-3-pyridinol
Hazard: Irritant
Formula:
C₁₁H₁₄ClNO₂Si MW: 255.78 g/mol Melting Pt: 96…98 °C |
MDL Number:
MFCD12827784 CAS Number: 208519-38-4 |
Specification Test Results
Melting point | 96-98°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...