2-Chloro-3-iodo-4-(trifluoromethyl)pyridine ≥90%
Supplier: Matrix Scientific
048516-500MG
048516-1G
048516-5G
101884-174EA
496.92
USD
101884-174
101884-176
101884-178
2-Chloro-3-iodo-4-(trifluoromethyl)pyridine ≥90%
2-Chloro-3-iodo-4-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₂ClF₃IN MW: 307.44 g/mol Melting Pt: 55…56 °C |
MDL Number:
MFCD12827836 CAS Number: 1186405-09-3 |
Specification Test Results
Melting point | 55-56°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...