2-Chloro-6-(dimethylamino)nicotinaldehyde ≥95%

Supplier: Matrix Scientific

048514-500MG 048514-1G
101884-164EA 269.23 USD
101884-164 101884-166
2-Chloro-6-(dimethylamino)nicotinaldehyde ≥95%
2-Chloro-6-(dimethylamino)nicotinaldehyde

Hazard: Irritant

Formula: C₈H₉ClN₂O
MW: 184.63 g/mol
Melting Pt: 111…112 °C
MDL Number: MFCD12827813
CAS Number: 1186405-06-0

Order Now

Specification Test Results

Melting point 111-112°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR