4-Chloro-2-(5-chloro-2-fluorophenyl)-5-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

048513-500MG 048513-1G 048513-5G
101884-158EA 747.69 USD
101884-158 101884-160 101884-162
4-Chloro-2-(5-chloro-2-fluorophenyl)-5-methoxypyrimidine ≥95%
4-Chloro-2-(5-chloro-2-fluorophenyl)-5-methoxypyrimidine

Hazard: Irritant

Formula: C₁₁H₇Cl₂FN₂O
Melting Pt: 128…130 °C
MDL Number: MFCD12827821
CAS Number: 674793-47-6

Order Now

Specification Test Results

Melting point 128-130°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR