2-Bromo-5,5-dimethyl-1,3-cyclohexanedione ≥95%

Supplier: Matrix Scientific

048510-500MG 048510-1G 048510-5G
101884-142EA 109.23 USD
101884-142 101884-144 101884-146
2-Bromo-5,5-dimethyl-1,3-cyclohexanedione ≥95%
2-Bromo-5,5-dimethyl-1,3-cyclohexanedione

Hazard: Irritant

Formula: C₈H₁₁BrO₂
Melting Pt: 158…160 °C
MDL Number: MFCD00092415
CAS Number: 1195-91-1

Order Now

Specification Test Results

Melting point 158-160°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR