6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Bromo-1,3-dihydro-2H-imidazo[4,5-b]pyridin-2-one
048509-500MG
048509-1G
101884-138EA
69.23
USD
101884-138
101884-140
6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one ≥95%
6-Bromo-1H-imidazo[4,5-b]pyridin-2(3H)-one
Hazard: Irritant
Formula:
C₆H₄BrN₃O MW: 214.0200 g/mol Melting Pt: >350 °C |
MDL Number:
MFCD00763285 CAS Number: 148038-83-9 |
Specification Test Results
Melting point | >350°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...