2-Bromo-1,3-cyclohexanedione ≥95%
Supplier: Matrix Scientific
048507-500MG
048507-1G
101884-132EA
133.85
USD
101884-132
101884-134
2-Bromo-1,3-cyclohexanedione ≥95%
2-Bromo-1,3-cyclohexanedione
Hazard: Irritant
Formula:
C₆H₇BrO₂ MW: 191.03 g/mol Melting Pt: 159…161 °C |
MDL Number:
MFCD00971951 CAS Number: 60060-44-8 |
Specification Test Results
Melting point | 159-161°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...