6-Bromo-2(3H)-benzoxazolone ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Bromo-2-benzoxazolinone, 6-Bromo-2-benzoxazolone, 6-Bromo-2-hydroxybenzoxazole, 6-Bromobenzo[d]oxazol-2(3H)-one
048506-500MG
048506-1G
048506-5G
101884-126EA
16.92
USD
101884-126
101884-128
101884-130
6-Bromo-2(3H)-benzoxazolone ≥95%
6-Bromo-2(3H)-benzoxazolone
Hazard: Irritant
Formula:
C₇H₄BrNO₂ MW: 214.018 g/mol Melting Pt: 193…195 °C |
MDL Number:
MFCD00694727 CAS Number: 19932-85-5 |
Specification Test Results
Melting point | 193-195°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...