3-(Benzyloxy)-N-methoxy-N-methyl-4-nitrobenzenecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Benzyloxy)-N-methoxy-N-methyl-4-nitrobenzamide

048503-500MG 048503-1G
101884-112EA 556.92 USD
101884-112 101884-114
3-(Benzyloxy)-N-methoxy-N-methyl-4-nitrobenzenecarboxamide ≥95%
3-(Benzyloxy)-N-methoxy-N-methyl-4-nitrobenzenecarboxamide

Hazard: Irritant

Formula: C₁₆H₁₆N₂O₅
MW: 316.32 g/mol
Melting Pt: 73…75 °C
MDL Number: MFCD12827824
CAS Number: 1186405-08-2

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR