[5-(Benzyloxy)-6-chloro-4-iodo-2-pyridinyl]methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Benzyloxy)-2-chloro-6-(hydroxymethyl)-4-iodopyridine

048502-500MG 048502-1G
101884-108EA 747.69 USD
101884-108 101884-110
[5-(Benzyloxy)-6-chloro-4-iodo-2-pyridinyl]methanol ≥95%
[5-(Benzyloxy)-6-chloro-4-iodo-2-pyridinyl]methanol

Hazard: Irritant

Formula: C₁₃H₁₁ClINO₂
Melting Pt: 122…124 °C
MDL Number: MFCD12827828
CAS Number: 1186405-17-3

Order Now

Specification Test Results

Melting point 122-124°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR