2-Amino-5-iodo-3-methylbenzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-iodo-3-methylbenzoic acid

048499-500MG 048499-1G 048499-5G
101884-098EA 110.77 USD
101884-098 101884-100 101884-102
2-Amino-5-iodo-3-methylbenzenecarboxylic acid ≥95%
2-Amino-5-iodo-3-methylbenzenecarboxylic acid

Hazard: Irritant

Formula: C₈H₈INO₂
MW: 277.06 g/mol
Melting Pt: 235…240 °C
MDL Number: MFCD09029816
CAS Number: 108857-24-5

Order Now

Specification Test Results

Melting point 235-240°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR