N-(2-Acetyl-4-iodo-6-methylphenyl)-2,2,2-trifluoroacetamide ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Acetyl-4-iodo-6-methyl-N-(trifluoroacetyl)aniline

048497-500MG 048497-1G 048497-5G
101884-090EA 375.38 USD
101884-090 101884-092 101884-094
N-(2-Acetyl-4-iodo-6-methylphenyl)-2,2,2-trifluoroacetamide ≥95%
N-(2-Acetyl-4-iodo-6-methylphenyl)-2,2,2-trifluoroacetamide

Hazard: Irritant

Formula: C₁₁H₉F₃INO₂
MW: 371.1 g/mol
Melting Pt: 136…138 °C
MDL Number: MFCD12827838
CAS Number: 935292-71-0

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR