Ethyl-2-(2-bromoethylidene)-1-hydrazinecarboxylate ≥95%

Supplier: Matrix Scientific

048227-500MG 048227-1G 048227-5G
101883-374EA 278.46 USD
101883-374 101883-376 101883-378
Ethyl-2-(2-bromoethylidene)-1-hydrazinecarboxylate ≥95%
Ethyl-2-(2-bromoethylidene)-1-hydrazinecarboxylate

Hazard: Irritant

Formula: C₅H₉BrN₂O₂
Melting Pt: 129 °C (dec.)
MDL Number: MFCD11841058
CAS Number: 62105-91-3

Order Now

Specification Test Results

Melting point 129°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR