4-Hydrazino-2-(methylsulfanyl)pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydrazinyl-2-(methylthio)pyrimidine

048225-500MG 048225-1G
101883-364EA 250.77 USD
101883-364 101883-366
4-Hydrazino-2-(methylsulfanyl)pyrimidine ≥95%
4-Hydrazino-2-(methylsulfanyl)pyrimidine

Hazard: Irritant

Formula: C₅H₈N₄S
MW: 156.21 g/mol
Melting Pt: 143…145 °C
MDL Number: MFCD01223746
CAS Number: 104408-29-9

Order Now

Specification Test Results

Melting point 143-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR