Methyl-5-bromo-2-(2-propynyloxy)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
048222-500MG
048222-1G
048222-5G
101883-352EA
356.92
USD
101883-352
101883-354
101883-356
Methyl-5-bromo-2-(2-propynyloxy)benzenecarboxylate ≥95%
Methyl-5-bromo-2-(2-propynyloxy)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₁H₉BrO₃ Melting Pt: 81…83 °C |
MDL Number:
MFCD11841056 CAS Number: 685126-90-3 |
Specification Test Results
Melting point | 81-83°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...