5-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

048219-500MG 048219-1G
101883-344EA 747.69 USD
101883-344 101883-346
5-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde ≥95%
5-(Trifluoromethyl)-1-benzothiophene-2-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₅F₃OS
Melting Pt: 66…68 °C
MDL Number: MFCD11841054
CAS Number: 1135283-83-8

Order Now

Specification Test Results

Melting point 66-68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR