2-Bromo-5-methoxy-4-methylaniline ≥95%

Supplier: Matrix Scientific

048218-500MG 048218-1G
101883-340EA 356.92 USD
101883-340 101883-342
2-Bromo-5-methoxy-4-methylaniline ≥95%
2-Bromo-5-methoxy-4-methylaniline

Hazard: Irritant

Formula: C₈H₁₀BrNO
MW: 216.0751 g/mol
Melting Pt: 75…77 °C
MDL Number: MFCD11841053
CAS Number: 532440-88-3

Order Now

Specification Test Results

Melting point 75-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR