(6-Chloro-1-benzothiophen-2-yl)methanol ≥95%
Supplier: Matrix Scientific
048212-500MG
048212-1G
101883-314EA
761.54
USD
101883-314
101883-316
(6-Chloro-1-benzothiophen-2-yl)methanol ≥95%
(6-Chloro-1-benzothiophen-2-yl)methanol
Hazard: Irritant
Formula:
C₉H₇ClOS Melting Pt: 111…113 °C |
MDL Number:
MFCD11841049 CAS Number: 234107-53-0 |
Specification Test Results
Melting point | 111-113°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...