(7-Fluoro-1-benzothiophen-2-yl)methanol ≥95%

Supplier: Matrix Scientific

Synonyms: 7-Fluoro-2-(hydroxymethyl)benzo[b]thiophene

048210-500MG 048210-1G
101883-308EA 356.92 USD
101883-308 101883-310
(7-Fluoro-1-benzothiophen-2-yl)methanol ≥95%
(7-Fluoro-1-benzothiophen-2-yl)methanol

Hazard: Irritant

Formula: C₉H₇FOS
MW: 182.22 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD11841047
CAS Number: 1135283-78-1

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR