(5-Fluoro-1-benzothiophen-2-yl)methanol ≥95%
Supplier: Matrix Scientific
048209-500MG
048209-1G
101883-304EA
375.38
USD
101883-304
101883-306
(5-Fluoro-1-benzothiophen-2-yl)methanol ≥95%
(5-Fluoro-1-benzothiophen-2-yl)methanol
Hazard: Irritant
Formula:
C₉H₇FOS Melting Pt: 91…93 °C |
MDL Number:
MFCD11841046 CAS Number: 13771-73-8 |
Specification Test Results
Melting point | 91-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...