6-Cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine ≥95%

Supplier: Matrix Scientific

048208-500MG 048208-1G
101883-300EA 556.92 USD
101883-300 101883-302
6-Cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine ≥95%
6-Cyclopropyl-1-methyl-1H-pyrazolo[3,4-b]pyridin-3-ylamine

Hazard: Irritant

Formula: C₁₀H₁₂N₄
Melting Pt: 142…145 °C
MDL Number: MFCD11841045
CAS Number: 1135283-77-0

Order Now

Specification Test Results

Melting point 142-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR