Methyl-5-(methylamino)-2-nitrobenzenecarboxylate ≥95%
Supplier: Matrix Scientific
048205-500MG
048205-1G
101883-290EA
278.46
USD
101883-290
101883-292
Methyl-5-(methylamino)-2-nitrobenzenecarboxylate ≥95%
Methyl-5-(methylamino)-2-nitrobenzenecarboxylate
Hazard: Irritant
Formula:
C₉H₁₀N₂O₄ Melting Pt: 134…136 °C |
MDL Number:
MFCD11841043 CAS Number: 1135283-73-6 |
Specification Test Results
Melting point | 134-136°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...