tert-Butyl-4-[2-(3-aminophenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%

Supplier: Matrix Scientific

048203-500MG 048203-1G
101883-286EA 556.92 USD
101883-286 101883-288
tert-Butyl-4-[2-(3-aminophenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate ≥95%
tert-Butyl-4-[2-(3-aminophenoxy)ethyl]tetrahydro-1(2H)-pyrazinecarboxylate

Hazard: Irritant

Formula: C₁₇H₂₇N₃O₃
MW: 321.42 g/mol
Melting Pt: 109…111 °C
MDL Number: MFCD11841042
CAS Number: 1135283-72-5

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR