3-(4-Ethylphenyl)-5-isoxazolamine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Amino-3-(4-ethylphenyl)isoxazole
048198-500MG
048198-1G
101883-266EA
556.92
USD
101883-266
101883-268
3-(4-Ethylphenyl)-5-isoxazolamine ≥95%
3-(4-Ethylphenyl)-5-isoxazolamine
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O MW: 188.23 g/mol Melting Pt: 109…111 °C |
MDL Number:
MFCD07382879 CAS Number: 1020955-20-7 |
Specification Test Results
Melting point | 109-111°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...