3,5-Dibromo-4-nitro-1H-pyrazole ≥95%
Supplier: Matrix Scientific
048187-1G
048187-5G
101883-226EA
146.15
USD
101883-226
101883-228
3,5-Dibromo-4-nitro-1H-pyrazole ≥95%
3,5-Dibromo-4-nitro-1H-pyrazole
Hazard: Irritant
Formula:
C3HBr2N3O2 MW: 270.8669 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD09972275 CAS Number: 104599-36-2 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...