2-[2-(Methylsulfanyl)phenoxy]acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: [2-(Methylthio)phenoxy]acetic acid

048184-500MG 048184-1G 048184-5G
101883-214EA 172.31 USD
101883-214 101883-216 101883-218
2-[2-(Methylsulfanyl)phenoxy]acetic acid ≥95%
2-[2-(Methylsulfanyl)phenoxy]acetic acid

Hazard: Irritant

Formula: C₉H₁₀O₃S
MW: 198.24 g/mol
Melting Pt: 167…168 °C
MDL Number: MFCD09972274
CAS Number: 3395-40-2

Order Now

Specification Test Results

Melting point 167-168°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR