N-Benzyl-6-chloro-2-methyl-4-pyrimidinamine ≥95%
Supplier: Matrix Scientific
048178-500MG
048178-1G
048178-5G
101883-186EA
181.54
USD
101883-186
101883-188
101883-190
N-Benzyl-6-chloro-2-methyl-4-pyrimidinamine ≥95%
N-Benzyl-6-chloro-2-methyl-4-pyrimidinamine
Hazard: Irritant
Formula:
C₁₂H₁₂ClN₃ Melting Pt: 63…69 °C |
MDL Number:
MFCD09972270 CAS Number: 1017783-05-9 |
Specification Test Results
Melting point | 63-69°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...