2-[(6-Chloro-2-methyl-4-pyrimidinyl)amino]-1-ethanol ≥95%
Supplier: Matrix Scientific
048177-500MG
048177-1G
048177-5G
101883-180EA
172.31
USD
101883-180
101883-182
101883-184
2-[(6-Chloro-2-methyl-4-pyrimidinyl)amino]-1-ethanol ≥95%
2-[(6-Chloro-2-methyl-4-pyrimidinyl)amino]-1-ethanol
Hazard: Irritant
Formula:
C₇H₁₀ClN₃O Melting Pt: 93…95 °C |
MDL Number:
MFCD09972269 CAS Number: 22177-97-5 |
Specification Test Results
Melting point | 93-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...